missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rg1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Ginsenoside Rg1 is one of the major active components of ginseng. Ginsenoside Rg1 ameliorates the impaired cognitive function, displays promising effects by reducing cerebral Aβ levels. Ginsenoside Rg1 also reduces NF-κB nuclear translocation.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rg1 |
| CAS | 22427-39-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C42H72O14 |
| Quantity | 10 mg |
| Synonym | Panaxoside A Panaxoside Rg1 |
| Solubility Information | DMSO : ≥ 100 mg/mL (124.84 mM) |
| SMILES | [H][C@]12C(C)(C)[C@@H](O)CC[C@]1(C)[C@]3([H])[C@@]([C@@](CC[C@]4([H])[C@@](CC/C=C(C)/C)(C)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)(C)[C@]4([H])[C@H](O)C3)(C)C[C@@H]2O[C@]6([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O |
| Molecular Weight (g/mol) | 801.01 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction