missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rd, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Ginsenoside Rd inhibits TNFα-induced NF-κB transcriptional activity with an IC50 of 12.05±0.82 μM in HepG2 cells. Ginsenoside Rd inhibits expression of COX-2 and iNOS mRNA. Ginsenoside Rd also inhibits Ca2+ influx. Ginsenoside Rd inhibits CYP2D6, CYP1A2, CYP3A4, and CYP2C9, with IC50s of 58.0±4.5 μM, 78.4±5.3 μM, 81.7±2.6 μM, and 85.1±9.1 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rd |
| CAS | 52705-93-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C48H82O18 |
| Quantity | 10 mg |
| Synonym | Gypenoside VIII |
| Solubility Information | DMSO : 100 mg/mL (105.58 mM; Need ultrasonic) |
| SMILES | C[C@](CC/C=C(C)/C)([C@]1([H])[C@]([C@H](O)C[C@@]2([H])[C@@]3(C)CC[C@]4([H])[C@]2(C)CC[C@H](O[C@]5([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@]6([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C4(C)C)([H])[C@@]3(C)CC1)O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O |
| Molecular Weight (g/mol) | 947.15 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction