missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rc, MedChemExpress
Description
Ginsenoside Rc, one of major Ginsenosides from Panax ginseng, enhances GABA receptorA (GABAA)-mediated ion channel currents (IGABA). Ginsenoside Rc inhibits the expression of TNF-α and IL-1β.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rc |
| CAS | 11021-14-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C53H90O22 |
| Quantity | 5 mg |
| Synonym | Panaxoside Rc |
| Solubility Information | DMSO : 100 mg/mL (92.66 mM; Need ultrasonic) ∣H2O : 50 mg/mL (46.33 mM; Need ultrasonic) |
| SMILES | C[C@](CC/C=C(C)/C)([C@]1([H])[C@]([C@H](O)C[C@@]2([H])[C@@]3(C)CC[C@]4([H])[C@]2(C)CC[C@H](O[C@]5([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@]6([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C4(C)C)([H])[C@@]3(C)CC1)O[C@@H]7O[C@H](CO[C@@H]8O[C@@H](CO)[C@H](O)[C@H]8O)[C@@H](O)[C@H](O)[C@H]7O |
| Molecular Weight (g/mol) | 1079.27 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?