missing translation for 'onlineSavingsMsg'
Learn More

Ginsenoside Rb2, MedChemExpress

Product Code. p-200096028
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16451278

Brand: Medchem Express HYN0040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ginsenoside Rb2 is one of the main bioactive components of ginseng extracts. Rb2 can upregulate GPR120 gene expression. Ginsenoside Rb2 has antiviral effects.

Ginsenoside Rb2 is one of the main bioactive components of ginseng extracts. Rb2 can upregulate GPR120 gene expression. Ginsenoside Rb2 has antiviral effects.

Specifications

Chemical Name or Material Ginsenoside Rb2
Physical Form Solid
CAS 11021-13-9
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C53H90O22
For Use With (Application) COVID-19-anti-virus
Synonym Ginsenoside C
Solubility Information H2O : ≥ 100 mg/mL (92.66 mM) ∣DMSO : 9.5 mg/mL (8.80 mM; Need ultrasonic and warming)
SMILES C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O[C@@](O[C@H](CO)[C@@H](O)[C@@H]4O)([H])[C@@H]4O[C@]([C@@H]([C@@H](O)[C@@H]5O)O)([H])O[C@@H]5CO)C)(CC[C@@]2([H])C3(C)C)[C@]6([C@@]1([H])[C@]([C@@](CC/C=C(C)/C)(C)O[C@@H]([C@@H]([C@@H](O)[C@@H]7O)O)O[C@@H]7CO[C@H](OC[C@H](O)[C@@H]8O)[C@@H]8O)([H])CC6)C
Molecular Weight (g/mol) 1079.27
Formula Weight 1079.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.