missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside C-K, MedChemExpress
Description
Ginsenoside C-K, a bacterial metabolite of G-Rb1, exhibits anti-inflammatory effects by reducing iNOS and COX-2. Ginsenoside C-K exhibits an inhibition against the activity of CYP2C9 and CYP2A6 in human liver microsomes with IC50s of 32.0±3.6 μM and 63.6±4.2 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Ginsenoside C-K |
| CAS | 39262-14-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H62O8 |
| Quantity | 10 mg |
| Synonym | Ginsenoside compound K Ginsenoside K |
| Solubility Information | DMSO : ≥ 100 mg/mL (160.55 mM) |
| SMILES | C[C@@]([C@@]12C)(CC[C@@]3([H])C4(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@]([C@@](CC/C=C(C)/C)(C)O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)([H])CC2)([H])[C@]3(CC[C@@H]4O)C |
| Molecular Weight (g/mol) | 622.87 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?