missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GINS4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30563
This item is not returnable.
View return policy
Description
GINS4 Polyclonal antibody specifically detects GINS4 in Human samples. It is validated for Western Blot
Specifications
| GINS4 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| DNA replication complex GINS protein SLD5, GINS complex subunit 4, GINS complex subunit 4 (Sld5 homolog), SLD5 homolog, SLD5MGC14799 | |
| Between 173 and 23 | |
| 100 μg | |
| DNA replication Transcription Translation and Splicing | |
| 84296 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction