missing translation for 'onlineSavingsMsg'
Learn More

GINS4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£342.00 - £470.00

Specifications

Antigen GINS4
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18467820
View Documents
Novus Biologicals
NBP1-82747-25ul
25 μL
£342.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18282538
View Documents
Novus Biologicals
NBP1-82747
0.1 mL
£470.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GINS4 Polyclonal specifically detects GINS4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

GINS4
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
DNA replication complex GINS protein SLD5, GINS complex subunit 4, GINS complex subunit 4 (Sld5 homolog), SLD5 homolog, SLD5MGC14799
GINS4
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
DNA replication Transcription Translation and Splicing
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
84296
This antibody was developed against Recombinant Protein corresponding to amino acids:VSIHQMEMERIRYVLSSYLRCRLMKIEKFFPHVLEKEKTRPEGEPSSLSPEELAFAREFMANTESYLK
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.