missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GINS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549424
This item is not returnable.
View return policy
Description
GINS1 Polyclonal Antibody for Western Blot
The yeast heterotetrameric GINS complex is made up of Sld5 (GINS4; MIM 610611), Psf1, Psf2 (GINS2; MIM 610609), and Psf3 (GINS3; MIM 610610). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
Specifications
| GINS1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9CZ15 | |
| GINS1 | |
| KLH conjugated synthetic peptide between 152-186 amino acids from the C-terminal region of Mouse Gins1. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GINS1 | |
| 2810418N01Rik; DNA replication complex GINS protein PSF1; GINS complex subunit 1; GINS complex subunit 1 (Psf1 homolog); GINS complex subunit 4 (Sld5 homolog); Gins1; Gins4; KIAA0186; mKIAA0186; partner of sld five-1; PSF1; RP4-691N24.2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 69270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction