missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GINS1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00009837-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MFCEKAMELIRELHRAPEGQLPAFNEDGLRQVLEEMKALYEQNQSDVNEAKSGGRSDLIPTIKFRHCSLLRNRRCTVAYLYDRLLRIRALRWEYGSILPNALRFHMAAEEMEWFNNYKRSLATYMRSLGGDEGLDITQDMKPPKSLYIEVRCLKDYGEFEVDDGTSVLLKKNSQHFLPRWKCEQLIRQGVLEHILSSpecifications
| GINS1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC012542 | |
| GINS1 | |
| GINS1 (AAH12542, 1 a.a. ∼ 196 a.a) full-length human protein. | |
| RUO | |
| 9837 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| GINS complex subunit 1 (Psf1 homolog) | |
| KIAA0186/PSF1 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion