missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolic Acid, MedChemExpress

Product Code. 16421298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421298 5 mg 5mg
16418858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16421298 Supplier Medchem Express Supplier No. HYN0077
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ginkgolic Acid is a natural compound that inhibits SUMOylation with an IC50 of 3.0 μM in in vitro assay.

Ginkgolic Acid is a natural compound that inhibits SUMOylation with an IC50 of 3.0 μM in in vitro assay.

Specifications

Chemical Name or Material Ginkgolic Acid
CAS 22910-60-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H34O3
Quantity 5 mg
Synonym Ginkgolic acid (15:1) Ginkgolic acid I Romanicardic acid
Solubility Information DMSO : 100 mg/mL (288.60 mM; Need ultrasonic)
SMILES O=C(O)C1=C(CCCCCCC/C=C\CCCCCC)C=CC=C1O
Molecular Weight (g/mol) 346.5
Formula Weight 346.5
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.