missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ginisortamab Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554901
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Ginisortamab targets and binds to specific antigens on cancer cells, facilitating immune system recognition and destruction of these cells.
Specifications
| Ginisortamab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CKTSF1B1; DAND2; DRM; GREM1; PIG2 | |
| Recombinant Human GREM1/DAND2. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1.57 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction