missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gimeracil, MedChemExpress
Click to view available options
Quantity:
25 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
50mg
Description
Gimeracil, a component of an oral fluoropyrimidine derivative S-1, inhibits DNA DSB repair and is a potent inhibitor of DPYD (dihydropyrimidine dehydrogenase, DPD).
Specifications
Specifications
| Chemical Name or Material | Gimeracil |
| Physical Form | Solid |
| CAS | 103766-25-2 |
| Color | White |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C5H4ClNO2 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | Gimestat |
| Solubility Information | DMSO : 100 mg/mL (687.10 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction