missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIMAP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11068-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.Specifications
| GIMAP5 | |
| Polyclonal | |
| Unconjugated | |
| GIMAP5 | |
| GIMAP5, GTPase IMAP family member 5, GTPase, IMAP family member 5, hIAN5, HIMAP3, IAN 5, IAN4, IAN4L1, IAN5, IMAP3, Immunity associated protein 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55340 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96F15 | |
| GIMAP5 | |
| GIMAP5 Fusion Protein Ag1509 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction