missing translation for 'onlineSavingsMsg'
Learn More

GIMAP5 Antibody (1E10), Novus Biologicals™

Product Code. 18420220 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420220 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420220 Supplier Novus Biologicals Supplier No. H00055340M10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIMAP5 Monoclonal antibody specifically detects GIMAP5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP5
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 1E10
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060854
Gene Alias FLJ11296, GTPase, IMAP family member 5, hIAN5, HIMAP3, IAN4, IAN4L1IROD, IAN5, IAN-5, IMAP3GTPase IMAP family member 5, immune associated nucleotide 4 like 1, immune associated nucleotide 4 like 1 (mouse), immunity associated protein 3, Immunity-associated nucleotide 4-like 1 protein, Immunity-associated nucleotide 5 protein, Immunity-associated protein 3, inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5
Host Species Mouse
Immunogen GIMAP5 (NP_060854.2, 186 a.a. ~ 284 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AFNNWGSVEEQRQQQAELLAVIERLGREREGSFHSNDLFLDAQLLQRTGAGACQEDYRQYQAKVEWQVEKHKQELRENESNWAYKALLRVKHLMLLHYE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 55340
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.