missing translation for 'onlineSavingsMsg'
Learn More

GIMAP4 Polyclonal Antibody

GIMAP4 Polyclonal Antibody

Brand:  Thermo Scientific PA555540

Product Code. 15743461

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GIMAP4 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GIMAP4
Polyclonal
Unconjugated
GIMAP4
AU019574; E430007K16Rik; GIMAP4; GTPase IMAP family member 4; GTPase, IMAP family member 4; human immune associated nucleotide 1; Ian1; IAN-1; Imap4; immune associated nucleotide 1; immune-associated nucleotide 1; immune-associated nucleotide-binding protein 1; immunity associated protein 4; immunity-associated nucleotide 1 protein; Immunity-associated protein 4; MSTP062
Rabbit
Antigen Affinity Chromatography
RUO
55303
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NUV9
GIMAP4
Recombinant protein corresponding to Human GIMAP4. Recombinant protein control fragment (Product #RP-92507).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.