missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIMAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101618
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GIMAP4.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The encoded protein of this gene may be negatively regulated by T-cell acute lymphocytic leukemia 1 (TAL1). In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications
| GIMAP4 | |
| Polyclonal | |
| Unconjugated | |
| GIMAP4 | |
| AU019574; E430007K16Rik; GIMAP4; GTPase IMAP family member 4; GTPase, IMAP family member 4; human immune associated nucleotide 1; Ian1; IAN-1; Imap4; immune associated nucleotide 1; immune-associated nucleotide 1; immune-associated nucleotide-binding protein 1; immunity associated protein 4; immunity-associated nucleotide 1 protein; Immunity-associated protein 4; MSTP062 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107526, 55303 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99JY3, Q9NUV9 | |
| GIMAP4 | |
| A synthesized peptide derived from human GIMAP4(Accession Q9NUV9), corresponding to amino acid residues H131-I181. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction