missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIMAP4 Monoclonal Antibody (OTI1C6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525993
This item is not returnable.
View return policy
Description
GIMAP4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The encoded protein of this gene may be negatively regulated by T-cell acute lymphocytic leukemia 1 (TAL1). In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications
| GIMAP4 | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NUV9 | |
| GIMAP4 | |
| Full length human recombinant protein of GIMAP4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1C6 | |
| Unconjugated | |
| GIMAP4 | |
| AU019574; E430007K16Rik; GIMAP4; GTPase IMAP family member 4; GTPase, IMAP family member 4; human immune associated nucleotide 1; Ian1; IAN-1; Imap4; immune associated nucleotide 1; immune-associated nucleotide 1; immune-associated nucleotide-binding protein 1; immunity associated protein 4; immunity-associated nucleotide 1 protein; Immunity-associated protein 4; MSTP062 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55303 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction