missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIMAP4 Antibody (OTI1C6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71997IR
This item is not returnable.
View return policy
Description
GIMAP4 Monoclonal specifically detects GIMAP4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GIMAP4 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GIMAP4 | |
| Full length recombinant protein of human GIMAP4 (NP_060796) produced in HEK293T cells. | |
| 0.1 mL | |
| 55303.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1C6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| GTPase IMAP family member 4, GTPase, IMAP family member 4, HIMAP4, human immune associated nucleotide 1, IAN-1, IAN1FLJ11110, IMAP4hIAN1, immunity associated protein 4, Immunity-associated nucleotide 1 protein, Immunity-associated protein 4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction