missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GIMAP4 Monoclonal antibody specifically detects GIMAP4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
Specifications
Specifications
| Antigen | GIMAP4 |
| Applications | Western Blot, Immunofluorescence, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI1A11 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | GTPase IMAP family member 4, GTPase, IMAP family member 4, HIMAP4, human immune associated nucleotide 1, IAN-1, IAN1FLJ11110, IMAP4hIAN1, immunity associated protein 4, Immunity-associated nucleotide 1 protein, Immunity-associated protein 4 |
| Host Species | Mouse |
| Immunogen | Full length recombinant protein of human GIMAP4 (NP_060796) produced in HEK293T cells. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?