missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIMAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101617
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GIMAP2.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications
| GIMAP2 | |
| Polyclonal | |
| Unconjugated | |
| GIMAP2 | |
| D15S9; GIMAP2; GTPase IMAP family member 2; GTPase, IMAP family member 2; HIMAP2; IAN12; IMAP2; immune-associated nucleotide-binding protein 12; immunity associated protein 2; immunity-associated protein 2; RNF63; ZNF127 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26157 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UG22 | |
| GIMAP2 | |
| A synthesized peptide derived from human GIMAP2(Accession Q9UG22), corresponding to amino acid residues E208-E258. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction