Learn More
GIMAP2 Mouse anti-Human, Unconjugated, Clone: 1F2A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67720-1-IG-20UL
This item is not returnable.
View return policy
Description
The GIMAP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GIMAP2. This antibody recognizes human antigen. The GIMAP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GIMAP2 | |
| Monoclonal | |
| 749 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1F2A2 | |
| Unconjugated | |
| 20 μL | |
| 26157 | |
| IgG1 |