missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIMAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560858
This item is not returnable.
View return policy
Description
Immunogen sequence: VCAFDNRATG REQEAQVEQL LGMVEGLVLE HKGAHYSNEV YELAQVLRWA GPEERLRRVA ERVAARVQ Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 65%.
GIMAP1 belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins.This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications
| GIMAP1 | |
| Polyclonal | |
| Unconjugated | |
| GIMAP1 | |
| GIMAP1; GTPase; GTPase IMAP family member 1; GTPase, IMAP family member 1; hIMAP1; Ian2; IAP38; imap; Imap1; Imap38; immune associated nucleotide family member; immune-associated nucleotide-binding protein 2; immune-associated protein 38; immunity associated protein 1; immunity-associated protein; immunity-associated protein 1; immunity-associated protein, 38 kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170575 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWP7 | |
| GIMAP1 | |
| Recombinant protein corresponding to Human GIMAP1. Recombinant protein control fragment (Product #RP-100002). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction