missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GILZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587243
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.
Specifications
| GILZ | |
| Polyclonal | |
| Unconjugated | |
| TSC22D3 | |
| delta sleep inducing peptide, immunoreactor; delta sleep-inducing peptide immunoreactor; DIP; Dsip1; DSIPI; DSIP-immunoreactive leucine zipper protein; DSIP-immunoreactive peptide; GILZ; glucocorticoid-induced leucine zipper protein; long glucocorticoid-induced leucine zipper protein; Protein DIP; Tilz3; TSC22 domain family 3; TSC22 domain family member 3; TSC22 domain family protein 3; TSC22 domain family, member 3; TSC-22 related protein; TSC22D3; TSC-22-like protein; TSC-22R; TSC22-related inducible leucine zipper 3; TSC-22-related protein; TSC22-related-inducible leucine zipper 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1831 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99576 | |
| TSC22D3 | |
| Recombinant full length Human TSC22D3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction