missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GilZ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38331-20ul
This item is not returnable.
View return policy
Description
GilZ Polyclonal antibody specifically detects GilZ in Human samples. It is validated for ELISA,Western Blot
Specifications
| GilZ | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| Delta sleep-inducing peptide immunoreactor, DSIP-immunoreactive leucine zipper protein, DSIP-immunoreactive peptide, GILZDKFZp313A1123, Glucocorticoid-induced leucine zipper protein, Protein DIP, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC-22 related protein, TSC-22-like protein, TSC-22-related protein, TSC-22Rimmunoreactor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 58-134 of human GilZ (NP_004080.2).,, Sequence:, MDLVKNHLMYAVREEVEILKEQIRELVEKNSQLERENTLLKTLASPEQLEKFQSCLSPEEPAPESPQVPEAPGGSAV | |
| 20 μL | |
| Cancer | |
| 1831 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction