missing translation for 'onlineSavingsMsg'
Learn More

GilZ Antibody (CFMKG15), DyLight 594, Novus Biologicals™

Product Code. 18431213 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431213 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431213 Supplier Novus Biologicals Supplier No. NBP200143DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

GilZ Monoclonal antibody specifically detects GilZ in Mouse samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GilZ
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone CFMKG15
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Delta sleep-inducing peptide immunoreactor, DSIP-immunoreactive leucine zipper protein, DSIP-immunoreactive peptide, GILZDKFZp313A1123, Glucocorticoid-induced leucine zipper protein, Protein DIP, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC-22 related protein, TSC-22-like protein, TSC-22-related protein, TSC-22Rimmunoreactor
Host Species Rat
Immunogen The immunogen for this antibody is GILZ.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1831
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.