Learn More
Invitrogen™ Gilvetmab Recombinant Canine Monoclonal Antibody
GREENER_CHOICE
Canine Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542149
Product Code. 17718773
  • £428.00 / 100µg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Gilvetmab is a biosimilar that targets PDCD1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gilvetmab
Recombinant Monoclonal
Unconjugated
AH-PD1
Canine (Canis lupus familiaris) PDCD1/PD1/CD279.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
3.4 mg/mL
PBS with no preservative; pH 7.4
Canine
Protein A/G
RUO
Canine
Antibody
IgG2 κ