missing translation for 'onlineSavingsMsg'
Learn More

Gilteritinib, MedChemExpress

Product Code. 16455866
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16455866 100 mg 100mg
16451387 50 mg 50mg
16405168 10 mg 10mg
16447887 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16455866 Supplier Medchem Express Supplier No. HY12432
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Gilteritinib (ASP2215) is a potent and ATP-competitive FLT3/AXL inhibitor with IC50s of 0.29 nM/0.73 nM, respectively.

Gilteritinib (ASP2215) is a potent and ATP-competitive FLT3/AXL inhibitor with IC50s of 0.29 nM/0.73 nM, respectively.

Specifications

Chemical Name or Material Gilteritinib
CAS 1254053-43-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H44N8O3
Quantity 100 mg
Synonym ASP2215
Solubility Information Ethanol : 100 mg/mL (180.93 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 2 mg/mL (3.62 mM; Need ultrasonic)
SMILES NC(C1=NC(CC)=C(NC2CCOCC2)N=C1NC3=CC(OC)=C(N4CCC(N5CCN(C)CC5)CC4)C=C3)=O
Molecular Weight (g/mol) 552.71
Formula Weight 552.71
Percent Purity 97.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.