missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Gilteritinib
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Gilteritinib is a potent Fms-like tyrosine kinase 3 (FLT3) and AXL inhibitor (IC50 = 0.29 nM and 0.73 nM, for FLT3 and AXL, respectively). In vivo, Gilteritinib reduces tumor growth and improves survival in xenograft and intra-bone marrow transplantation models of FLT3 mutated acute myeloid leukemia. Gilteritinib is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 6-Ethyl-3-[[3-methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]amino]-5-[(tetrahydro-2H-pyran-4-yl)amino]-2-pyrazinecarboxamide |
| CAS | 1254053-43-4 |
| Quantity | 50 mg |
| Target | Flt-3/Flk-2/CD135 |
| Molecular Formula | C29H44N8O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction