Learn More
Gilson™ Universal Multichannel Stand
Universal Multichannel Stand designed to accomodate 8 or 12 channel pipets.
Brand: Gilson™ F161417
Code : 50
Additional Details : Weight : 0.14400kg
Description
Universal Multichannel Stand is made of Altuglas™ Essential quality PMMA (Polymethyl Methacrylate). PMMA is highly transparent and offers a good resistance to scratches, impacts and temperature. It also has a good resistance to many chemicals commonly handled in laboratories such as alcohols like ethanol and methanol, disinfectants and some acids and bases depending on the concentration.
- Made of flexible plastic
- Support edges widen or retract to adapt geometry of virtually all pipet brands
Specifications
Clear | |
Pipettes | |
PMMA | |
All Gilson and other brands of multichannel pipet |