missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MICROMAN™ Positive-Displacement Pipets

Product Code. 10585254 Shop All Gilson Products
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
100 to 1000 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
M10
M100
M1000
M25
M250
M50
Compatible Tips:
C10
C100
C1000
C25
C250
C50
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10585254

Brand: Gilson™ F148502

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ensures accuracy and precision when pipetting problem liquids, and offers full protection against cross contamination. Gilson™ MICROMAN™ Positive-Displacement Pipets are 3 to 4 times better than an air displacement pipet when pipetting viscous or volatile liquids.

  • Piston is located inside the tip or capillary preventing pipet contamination
  • Color-coded push-button allows for easy identification of different models
  • Dual function push-button that also ejects capillary pistons
  • Comfortable PVDF handle resistant to aggressive chemicals
  • Accurately measure high vapor pressure liquids without leakage or degradation of the tip
  • M10 and M100 models are fully autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips C25
Autoclavable Autoclavable
Model M25
Number of Channels 1
Type Positive Displacement Pipette
Volume (Metric) 3 to 25 μL
For Use With (Application) Pipetting of viscous and dense liquids
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.