Learn More
Innocal™ NIST-Traceable Calibration for Peristaltic Pump Drive
NIST-Traceable Calibration report with data for Peristaltic Pump Drive.
Brand:  Innocal™ 17050-01
Code : A0
Additional Details : Weight : 0.04000kg
Product Code. 13218169
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards.
  • Ensure precision in your process.
  • More accurate volumes, better system control.
  • Technicians can calibrate a peristaltic pump drive to ensure accurate rotation per minute (rpm) output.
Specifications
Specifications
Peristaltic Pump Drive
NIST-Traceable Calibration
Ensure precision in your process.