Learn More
Epredia™ Gill™ Hematoxylin 1, 2, and 3
Shop All Epredia Products
Click to view available options
Quantity:
1 pt.
1 gal.
Type:
Gill 1 Hematoxylin
Gill 2 Hematoxylin
Gill 3 Hematoxylin
Unit Size:
Each
Pack of 4
Description
Gill Hematoxylins are progressive nuclear stains.
- Gill 1 is recommended for use in cytology
- Gill 2 has applications in histology and cytology
- Gill 3 is used in histology
- All are pre-mixed and ready-to-use
Specifications
Specifications
| Type | Gill 3 Hematoxylin |
| Quantity | 1 gal. |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H371-May cause damage to organs.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : aluminium sulphate,sodium iodate, hematoxylin, ethylene glycol, diazolidinyl urea, acetic acid