missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Gilactiv™ Container with Lid
Description
- Proven antibacterial and antiviral effectiveness on the container's surface (empty or full) for a broad spectrum of germs, including HCoV-229E human coronavirus.
- This article contains a biocidal product with the active substance: Silver Phosphate Glass.
- Tests conducted by an EU-approved French independent laboratory showed that 98.6% of the virus was wiped out after a two-hour period.
- Choose Gilactiv™ enhanced protection.
- Test results available on request.
- No gaps nor overlaps and no sharp corners.
- Rewritable identification cartridge for inventory tracking.
- Recessed handles for better handling.
- Smooth interior walls and rounded corners for easier cleaning.
- Machine washable.
- Compatible with all GILAC 4 and 6-wheel dollies.
Specifications
Specifications
| Type | Storage Tote |
| Material | HDPE |
| Color | Blue |
| Shape | Rectangular |
| Capacity (English) | 4 gal. |
| Capacity (Metric) | 15 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?