missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIGYF2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GIGYF2 |
|---|---|
| Concentration | 0.26 mg/mL |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16883285
|
Proteintech
24790-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16873285
|
Proteintech
24790-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function of this protein remains unknown.Specifications
| GIGYF2 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q6Y7W6 | |
| 26058 | |
| GIGYF2 Fusion Protein Ag20329 | |
| Primary | |
| -20°C | |
| GIGYF2 |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GIGYF2, GYF2, KIAA0642, PARK11, PERQ2, PERQ3, TNRC15 | |
| GIGYF2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title