missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GIGYF2 Polyclonal Antibody

Product Code. 15771562 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15771562 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15771562 Supplier Invitrogen™ Supplier No. PA563793

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QQYAQVLAQQ QKAALSSQQQ QQLALLLQQF QTLKMRISDQ NIIPSVTRSV SVPDTGSIWE LQPTASQPTV WEGGSVWDLP L Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIGYF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GIGYF2
Gene Accession No. Q6Y7W6
Gene Alias 2610016F01Rik; A830080H02Rik; AI852361; AW259676; BC006835; GIGYF2; GRB10 interacting GYF protein 2; GRB10 interacting GYF protein 2; GRB10-interacting GYF protein 2; GRB10-interacting GYF protein 2; GYF2; KIAA0642; LOW QUALITY PROTEIN: PERQ amino acid-rich with GYF domain-containing protein 2; mKIAA0642; PARK11; Parkinson disease (autosomal recessive, early onset) 11; PERQ amino acid rich, with GYF domain 3; PERQ amino acid-rich with GYF domain-containing 2-like protein; PERQ amino acid-rich with GYF domain-containing protein 2; PERQ amino acid-rich with GYF domain-containing protein 2; GRB10-interacting GYF protein 2; PERQ2; PERQ3; rCG23949-like; TNRC15; trinucleotide repeat containing 15; trinucleotide repeat-containing gene 15 protein
Gene Symbols GIGYF2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human GIGYF2 (Product #RP-99707)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.