missing translation for 'onlineSavingsMsg'
Learn More

Gigaxonin Antibody, DyLight 350, Novus Biologicals™

Product Code. 18412243 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412243 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412243 Supplier Novus Biologicals Supplier No. NBP149923UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gigaxonin Polyclonal antibody specifically detects Gigaxonin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gigaxonin
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ38059, GAN1KLHL16Kelch-like protein 16, giant axonal neuropathy (gigaxonin), gigaxonin
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human Gigaxonin using the numbering given in entry NP_071324.1 (GeneID 8139).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8139
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.