missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gigaxonin Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49923APC
This item is not returnable.
View return policy
Description
Gigaxonin Polyclonal antibody specifically detects Gigaxonin in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| Gigaxonin | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| FLJ38059, GAN1KLHL16Kelch-like protein 16, giant axonal neuropathy (gigaxonin), gigaxonin | |
| The immunogen for this product maps to a region between residue 50 and 100 of human Gigaxonin using the numbering given in entry NP_071324.1 (GeneID 8139). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8139 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction