missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GIF Recombinant Rabbit Monoclonal Antibody (016)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA530462

Product Code. 16393994

  • £408.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Gastric intrinsic factor/GIF.

Promotes absorption of the essential vitamin cobalamin (Cbl) in the ileum. After interaction with CUBN, the GIF-cobalamin complex is internalized via receptor-mediated endocytosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GIF
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P27352
CBLIF
Recombinant Human Gastric intrinsic factor/GIF Protein (Met1-Tyr417).
100 μL
Primary
Human
Antibody
IgG
ELISA
016
Unconjugated
CBLIF
AV073125; Cblif; Cobalamin binding intrinsic factor; gastric intrinsic factor; gastric intrinsic factor (vitamin B synthesis); gastric intrinsic factor-like; Gif; IF; IFMH; INF; intestinal cobalamin binding protein; intrinsic factor; Pancreatic intrinsic factor; TCN3
Rabbit
Protein A
RUO
2694
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.