missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIF Rabbit anti-Human, Clone: 12, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90226-50UL
This item is not returnable.
View return policy
Description
GIF Monoclonal specifically detects GIF in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| GIF | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2694 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 12 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate | |
| gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3 | |
| Recombinant Human GIF Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction