missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587282
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Promotes absorption of the essential vitamin cobalamin (Cbl) in the ileum. After interaction with CUBN, the GIF-cobalamin complex is internalized via receptor-mediated endocytosis.
Specifications
| GIF | |
| Polyclonal | |
| Unconjugated | |
| CBLIF | |
| AV073125; Cblif; Cobalamin binding intrinsic factor; gastric intrinsic factor; gastric intrinsic factor (vitamin B synthesis); gastric intrinsic factor-like; Gif; IF; IFMH; INF; intestinal cobalamin binding protein; intrinsic factor; Pancreatic intrinsic factor; TCN3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14603, 2694 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P27352, P52787 | |
| CBLIF | |
| Recombinant full length Human GIF. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction