missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569550
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
Promotes absorption of the essential vitamin cobalamin (Cbl) in the ileum. After interaction with CUBN, the GIF-cobalamin complex is internalized via receptor-mediated endocytosis.
Specifications
| GIF | |
| Polyclonal | |
| Unconjugated | |
| CBLIF | |
| AV073125; Cblif; Cobalamin binding intrinsic factor; gastric intrinsic factor; gastric intrinsic factor (vitamin B synthesis); gastric intrinsic factor-like; Gif; IF; IFMH; INF; intestinal cobalamin binding protein; intrinsic factor; Pancreatic intrinsic factor; TCN3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14603 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P52787 | |
| CBLIF | |
| synthetic peptide directed towards the C-terminal region of mouse GIF. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction