missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIF Antibody (016), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90227CL1
This item is not returnable.
View return policy
Description
GIF Monoclonal antibody specifically detects GIF in Human samples. It is validated for ELISA,Sandwich ELISA Capture
Specifications
| GIF | |
| Monoclonal | |
| CoraFluor 1 | |
| gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human GIF (Uniprot#: P27352-1; Met1-Tyr417). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA Capture | |
| 016 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2694 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction