missing translation for 'onlineSavingsMsg'
Learn More

GIF Antibody (007), Biotin, Novus Biologicals™

Product Code. 18343740 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343740 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343740 Supplier Novus Biologicals Supplier No. NBP290225B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GIF Monoclonal specifically detects GIF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIF
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 7
Conjugate Biotin
Formulation PBS
Gene Alias gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human GIF (P27352-1; Met1-Tyr417).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2694
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.