missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GIF Monoclonal specifically detects GIF in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | GIF |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 7 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human GIF (P27352-1; Met1-Tyr417). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?