missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Giemsa Stain, pure, high purity biological stain

Product Code. 10736001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10736001 5 g Glass bottle 5g
10696542 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10736001 Supplier Thermo Scientific Chemicals Supplier No. 153230050
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 51811-82-6 | C14H14ClN3S | 291.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.80
MDL Number MFCD00012112,MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1

Specifications

Absorbance 950 min. (in MeOH, on dried substance), 600 min. (in MeOH, on dried substance) A (1%/1cm)
Melting Point 300.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
Lambda Max 640 to 650nm (in MeOH), 520 to 525nm (in MeOH)
Loss on Drying 10% max. (110°C)
Grade Pure
Packaging Glass bottle
Color Black to Green
Quantity 5 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Giemsa Stain
Show More Show Less
compliance-icons
Product Identifier
  • Giemsa Stain
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.