missing translation for 'onlineSavingsMsg'
Learn More

Giemsa Stain (Crystalline), Fisher BioReagents

Product Code. p-26049
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass Bottle
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 10002764

Brand: Fisher BioReagents BP24225

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Giemsa is a biological stain used for thin blood films to differentiate leukocytes, for staining malarial parasites in thin and thick blood films, and for staining bone marrow to show cell morphology.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.80
MDL Number MFCD00012112,MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1

Specifications

Melting Point 300°C
Bio. Stain Comm. Certification Pass Test
Identification Pass Test
Formula Weight 291.8
Packaging Glass Bottle
Color Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Giemsa Stain
compliance-icons
Product Identifier
  • Giemsa Stain
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.