Learn More
Epredia™ Giemsa Stain
Product Code. 12583687 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Unit Size:
Each
Product Code. 12583687
Brand: Epredia™ 89004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Take advantage of an excellent classical blood film stain used on peripheral blood smears and bone marrow with the Epredia™ Giemsa Stain.
  • Erythrocytes stain pink with platelets exhibiting a very light, pale hue
  • Leukocyte nuclear chromatin stains magenta-purple
  • Monocyte cytoplasm stains the classical “ground glass” blue
  • Lymphocyte cytoplasm stains clear, sky blue
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 gal.
Type Giemsa Stain
compliance-icons compliance-icons compliance-icons
Product Identifier
  • STAIN,GIEMSA,1GL CUBE
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H370-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : Methanol