missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GID4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569987
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The multiprotein Mediator complex is a coactivator required for activation of RNA polymerase II transcription by DNA bound transcription factors. The protein encoded by this gene is thought to be a subunit of the Mediator complex. This gene is located within the Smith-Magenis syndrome region on chromosome 17.
Specifications
| GID4 | |
| Polyclonal | |
| Unconjugated | |
| GID4 | |
| 4933439F18Rik; C17orf39; C19H17orf39; C5H17orf39; GID complex subunit 4; GID complex subunit 4 homolog; GID complex subunit 4 homolog; glucose-induced degradation protein 4 homolog; GID complex subunit 4, VID24 homolog; GID complex subunit 4, VID24 homolog (S. cerevisiae); GID4; Glucose-induced degradation protein 4 homolog; im:7147459; LOC509503 protein; LOW QUALITY PROTEIN: glucose-induced degradation protein 4 homolog; vacuolar import and degradation 24; vacuolar import and degradation protein 24 homolog; VID24 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79018 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IVV7 | |
| GID4 | |
| synthetic peptide directed towards the C terminal region of human GID4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction