missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Viral Production Cells

Product Code. p-7113084
Click to view available options
Quantity:
1 mL
6 x 1 mL
Unit Size:
1mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15967871

Brand: Gibco™ A35347

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Viral Production Cells are a derivative of the HEK 293F cell line and are a core component of the LV-MAX Lentiviral Production System.

Gibco Viral Production Cells are a derivative of the HEK 293F cell line and are a core component of the LV-MAX Lentiviral Production System (Cat. No. A35684). Viral Production Cells have been adapted to a special chemically-defined, serum-free and protein-free LV-MAX Production Medium (A3583401). They are maintained in suspension culture and will grow to high density in LV-MAX Production Medium. Viral Production Cells are highly transfectable by LV-MAX Transfection reagent (A35348) and generate superior lentiviral production compared to polyethyleneimine (PEI)-based lentiviral production methods.

  • Suspension culture cells facilitate high-throughput screening or scale-up
  • Higher viral production levels than other suspension HEK 293 cells
  • Rapid cell growth and high culture viabilities

Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in LV-MAX Production Medium and 10% DMSO. The cells should be thawed directly into LV-MAX Production Medium.

The cells are typically grown in flasks on a shaker platform in a humidified, CO2 cell culture incubator. However, cultures are scalable in volume from 96-deep well blocks to 2-L flasks and bioreactors. Lentiviral vector harvest is typically performed at two days post-transfection.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen until ready to use. Do not store the cells at -80°C.
Cell Line Viral Production Cell-Line
Cell Type Kidney (Embryonic)
Culture Type Suspension Cell Culture
Media Recommendation LV-MAX™ Production Medium
No. of Cells 1 x 107
Product Type Viral Production Cells
Quantity 1 mL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.