Learn More
Gibco™ Trypan Blue Solution, 0.4%

Description
- Performed while counting cells with hemocytometer during routine subculturing, but can be performed any time cell viability needs to be determined quickly and accurately
- Dye exclusion test is based upon concept that viable cells do not take up impermeable dyes (like Trypan Blue), but dead cells are permeable and take up the dye
- Produced at a cGMP-compliant facility located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Blue |
| Content And Storage | Storage conditions: 15 – 30°C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
| Cell Type | Mammalian Cells |
| Concentration | 0.004 |
| Detection Method | Colorimetric |
| Form | Liquid |
| Product Type | Trypan Blue |
| Sterility | Sterile-filtered |
| For Use With (Application) | Cell Counting |
| Quantity | 100 mL |
| Show More |
Frequently Asked Questions (FAQs)
There are a number of reagents used to quench fluorescence on the outside of live cells or in the media (background fluorescence). Our Backdrop Background Suppressor reagents are a great option to quench blue, green, and red fluorescent dyes, various media components and fluorescent proteins. Another commonly used reagent is the cell impermeant colorimetric dye, Trypan blue.
Trypan Blue is filter sterilized through a 0.22 µm filter.
It is a 0.85% saline solution.
Unfortunately, there is no definitive answer to this question. The rate at which the cell-impermeant dye is absorbed depends on the cell type, their state of health, nourishment, engulfment activity, etc.
Exposure to light may degrade the dye and these contaminants may promote precipitation. Trypan blue can also form orange/red fibrous aggregates if exposed to refrigeration or freezing temperatures.
Safety and Handling
- TRYPAN BLUE STAIN
- Danger
- Carcinogenicity Category 1
- Reproductive toxicity Category 2
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 2,7-Naphthalenedisulfonic acid, 3,3-[(3,3-dimethyl[1,1-biphenyl]-4,4-diyl)bis(azo)]bis[5-amino-4-hyd
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.